Assay Design



Gene Transcripts

Gene Symbol
PWD
Entrez Gene ID
828547
Gene Name
phosphoglucan, water dikinase
Gene Aliases
AT4G24450, ATGWD2, GWD3, T22A6.280, T22A6_280, phosphoglucan, water dikinase
Chromosome Location
Chr.4: 12635061 - 12642808 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341673.1 NP_001328878.1 23 - 24 2870 114
NM_001341674.1 NP_001328879.1 23 - 24 2874 114
NM_118578.5 NP_194176.3 22 - 23 2672 114

View Full Product Details