Assay Design



Gene Transcripts

Gene Symbol
IPT4
Entrez Gene ID
828567
Gene Name
isopentenyltransferase 4
Gene Aliases
ARABIDOPSIS THALIANA ISOPENTENYLTRANSFERASE 4, AT4G24650, ATIPT4, F22K18.150, F22K18_150, isopentenyltransferase 4
Chromosome Location
Chr.4: 12720394 - 12721443 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118598.1 NP_194196.1 1 - 1 941 87
GenBank mRNA AB061402.1 - 1 - 1 941 87
AB062611.1 - 1 - 1 869 87

View Full Product Details