Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001341698.1 | NP_001329102.1 | 6 - 7 | 723 | 74 |
NM_001341699.1 | NP_001329107.1 | 9 - 10 | 1123 | 74 | |
NM_001341701.1 | NP_001329104.1 | 7 - 8 | 1874 | 74 | |
NM_001341702.1 | NP_001329105.1 | 7 - 8 | 1874 | 74 | |
NM_001341703.1 | NP_001329106.1 | 8 - 9 | 1729 | 74 | |
NM_001341704.1 | NP_001329108.1 | 5 - 6 | 1710 | 74 | |
NM_001341705.1 | NP_001329109.1 | 8 - 9 | 1257 | 74 | |
NM_118607.3 | NP_194205.1 | 9 - 10 | 1157 | 74 | |
NM_202881.2 | NP_974610.1 | 8 - 9 | 967 | 74 | |
GenBank mRNA | AK227055.1 | - | 9 - 10 | 1029 | 74 |
BT025991.1 | - | 9 - 10 | 944 | 74 | |
D45353.1 | - | 9 - 10 | 1064 | 74 | |
U16177.1 | - | 9 - 10 | 987 | 74 |