Assay Design
Gene Transcripts
- Gene Symbol
- PAP24
- Entrez Gene ID
-
828591
- Gene Name
- purple acid phosphatase 24
- Gene Aliases
-
ARABIDOPSIS THALIANA PURPLE ACID PHOSPHATASE 24, AT4G24890, ATPAP24, F13M23.30, F13M23_30, purple acid phosphatase 24
- Chromosome Location
-
Chr.4: 12811354 - 12814456 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_118621.4 |
NP_194219.2 |
7 - 8 |
780 |
84 |
GenBank mRNA |
AY142520.1 |
- |
7 - 8 |
764 |
84 |
View Full Product Details