Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_118697.4 | NP_567725.1 | 6 - 7 | 1468 | 87 |
NM_179113.2 | NP_849444.1 | 5 - 6 | 1474 | 87 | |
GenBank mRNA | AF424575.1 | - | 6 - 7 | 1374 | 87 |
AY056168.1 | - | 6 - 7 | 1397 | 87 | |
AY062731.1 | - | 5 - 6 | 1474 | 87 | |
AY344062.1 | - | 6 - 7 | 1361 | 87 | |
BT002586.1 | - | 5 - 6 | 1430 | 87 |