Assay Design
Gene Transcripts
- Gene Symbol
- GPP1
- Entrez Gene ID
-
828690
- Gene Name
- glycerol-3-phosphatase 1
- Gene Aliases
-
AT4G25840, F14M19.120, F14M19_120, glycerol-3-phosphatase 1
- Chromosome Location
-
Chr.4: 13138957 - 13141097 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_118717.3 |
NP_567731.1 |
3 - 4 |
604 |
98 |
View Full Product Details