Assay Design



Gene Transcripts

Gene Symbol
CIL
Entrez Gene ID
828705
Gene Name
CCT motif family protein
Gene Aliases
AT4G25990, F20B18.100, F20B18_100
Chromosome Location
Chr.4: 13191085 - 13194124 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001341771.1 NP_001328403.1 1 - 2 1559 111
NM_118732.2 NP_567737.1 1 - 2 1559 111
NM_179114.2 NP_849445.1 1 - 2 1055 111
GenBank mRNA AB493701.1 - 1 - 2 978 111
AF359388.1 - 1 - 2 978 111
BT030317.1 - 1 - 2 978 111

View Full Product Details