Assay Design
Gene Transcripts
- Gene Symbol
- CGA1
- Entrez Gene ID
-
828721
- Gene Name
- cytokinin-responsive gata factor 1
- Gene Aliases
-
AT4G26150, F20B18.260, F20B18_260, GATA TRANSCRIPTION FACTOR 22, GATA22, GNC-LIKE, GNL, cytokinin-responsive gata factor 1
- Chromosome Location
-
Chr.4: 13253084 - 13255106 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_118748.3 |
NP_194345.1 |
2 - 3 |
782 |
98 |
View Full Product Details