Assay Design
Gene Transcripts
- Gene Symbol
- ACHT1
- Entrez Gene ID
-
828722
- Gene Name
- atypical CYS HIS rich thioredoxin 1
- Gene Aliases
-
AT4G26160, atypical CYS HIS rich thioredoxin 1, atypical CYS HIS rich thioredoxin 1
- Chromosome Location
-
Chr.4: 13255251 - 13256879 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_118749.4 |
NP_194346.1 |
1 - 2 |
269 |
121 |
GenBank mRNA |
BT024608.1 |
- |
1 - 2 |
224 |
121 |
View Full Product Details