Assay Design



Gene Transcripts

Gene Symbol
MAP65-2
Entrez Gene ID
828783
Gene Name
microtubule-associated protein 65-2
Gene Aliases
AT4G26760, F10M23.100, F10M23_100, microtubule-associated protein 65-2
Chromosome Location
Chr.4: 13478592 - 13481870 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118810.4 NP_567756.1 3 - 4 516 112
GenBank mRNA AY085430.1 - 3 - 4 397 112
BT030366.1 - 2 - 3 272 112

View Full Product Details