Assay Design



Gene Transcripts

Gene Symbol
TOR1
Entrez Gene ID
828814
Gene Name
ARM repeat superfamily protein
Gene Aliases
AT4G27060, CN, CONVOLUTA, SPIRAL 2, SPR2, T24A18.10, T24A18_10, TORTIFOLIA 1
Chromosome Location
Chr.4: 13581286 - 13585250 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_118840.5 NP_194436.2 1 - 2 900 62
GenBank mRNA AB176521.1 - 1 - 2 720 62
AJ249836.2 - 1 - 2 799 62

View Full Product Details