Assay Design
Gene Transcripts
- Gene Symbol
- VIIIB
- Entrez Gene ID
-
828845
- Gene Name
- P-loop containing nucleoside triphosphate hydrolases superfamily protein
- Gene Aliases
-
AT4G27370, ATVIIIB, F27G19.14, MYOSIN VIII B
- Chromosome Location
-
Chr.4: 13694689 - 13701119 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_118871.6 |
NP_194467.5 |
2 - 3 |
485 |
122 |
View Full Product Details