Assay Design



Gene Transcripts

Gene Symbol
CIP7
Entrez Gene ID
828851
Gene Name
COP1-interacting protein 7
Gene Aliases
AT4G27430, COP1-interacting protein 7, F27G19.30, F27G19_30
Chromosome Location
Chr.4: 13718007 - 13723461 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125596.2 NP_001119068.1 5 - 6 591 73
NM_118877.4 NP_194473.1 5 - 6 1263 73
GenBank mRNA AB012912.1 - 5 - 6 591 73

View Full Product Details