Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_112002.3 | NP_566398.1 | 1 - 2 | 1655 | 78 |
GenBank mRNA | AY056197.1 | - | 1 - 2 | 1467 | 78 |
AY058879.1 | - | 1 - 2 | 1474 | 78 | |
AY133794.1 | - | 1 - 2 | 1321 | 78 |