Assay Design



Gene Transcripts

Gene Symbol
NF-YC11
Entrez Gene ID
820427
Gene Name
nuclear factor Y, subunit C11
Gene Aliases
''nuclear factor Y, AT3G12480, nuclear factor Y, subunit C11, subunit C11''
Chromosome Location
Chr.3: 3957763 - 3960472 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112083.4 NP_187854.2 5 - 6 690 71
GenBank mRNA AF462841.1 - 5 - 6 616 71
AY143847.1 - 5 - 6 388 71

View Full Product Details