Assay Design



Gene Transcripts

Gene Symbol
PSAE-1
Entrez Gene ID
828996
Gene Name
Photosystem I reaction centre subunit IV / PsaE protein
Gene Aliases
AT4G28750, PSA E1 KNOCKOUT, PSI-E
Chromosome Location
Chr.4: 14202758 - 14204061 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119019.4 NP_567818.2 2 - 3 495 64
GenBank mRNA AJ245908.1 - 2 - 3 388 64
AY042790.1 - 2 - 3 389 64
AY081687.1 - 2 - 3 322 64

View Full Product Details