Assay Design



Gene Transcripts

Gene Symbol
CNGC17
Entrez Gene ID
829159
Gene Name
cyclic nucleotide-gated channel 17
Gene Aliases
AT4G30360, ATCNGC17, CYCLIC NUCLEOTIDE-GATED CHANNEL 17, F17I23.300, F17I23_300, cyclic nucleotide-gated channel 17
Chromosome Location
Chr.4: 14854694 - 14858019 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119182.4 NP_194765.2 4 - 5 1436 64
GenBank mRNA AK227176.1 - 4 - 5 1413 64
AY124000.1 - 4 - 5 1412 64
BT002284.1 - 4 - 5 1196 64

View Full Product Details