Assay Design



Gene Transcripts

Gene Symbol
ATG18B
Entrez Gene ID
829174
Gene Name
autophagy 18 B-like protein
Gene Aliases
ARABIDOPSIS THALIANA HOMOLOG OF YEAST AUTOPHAGY 18 (ATG18) B, AT4G30510, ATATG18B, F17I23.150, F17I23_150, homolog of yeast autophagy 18 (ATG18) B
Chromosome Location
Chr.4: 14905097 - 14907645 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342010.1 NP_001320091.1 10 - 11 1032 133
NM_001342011.1 NP_001320092.1 8 - 9 1216 133
GenBank mRNA AY142529.1 - 8 - 9 818 133

View Full Product Details