Assay Design
Gene Transcripts
- Gene Symbol
- ATS2
- Entrez Gene ID
-
829181
- Gene Name
- Phospholipid/glycerol acyltransferase family protein
- Gene Aliases
-
AT4G30580, EMB1995, EMBRYO DEFECTIVE 1995, F17I23.80, F17I23_80, LPAT1, PHOSPHOLIPID/GLYCEROL ACYLTRANSFERASE, lysophosphatidic acid acyltransferase 1
- Chromosome Location
-
Chr.4: 14932105 - 14934528 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_119204.4 |
NP_194787.2 |
5 - 6 |
792 |
118 |
GenBank mRNA |
AK118028.1 |
- |
5 - 6 |
783 |
118 |
View Full Product Details