Assay Design



Gene Transcripts

Gene Symbol
NFD1
Entrez Gene ID
829217
Gene Name
Ribosomal protein L21
Gene Aliases
AT4G30925, AT4G30930, NUCLEAR FUSION DEFECTIVE 1
Chromosome Location
Chr.4: 15049811 - 15051751 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119240.4 NP_567861.1 4 - 5 812 151
GenBank mRNA AB493710.1 - 4 - 5 691 151
AK228212.1 - 4 - 5 721 151
AY088563.1 - 4 - 5 726 151
BT005870.1 - 4 - 5 691 151

View Full Product Details