Assay Design



Gene Transcripts

Gene Symbol
SKB1
Entrez Gene ID
829240
Gene Name
SHK1 binding protein 1
Gene Aliases
AT4G31120, ATPRMT5, CAU1, F6E21.40, F6E21_40, PRMT5, PROTEIN ARGININE METHYLTRANSFERASE 5, SHK1 binding protein 1, calcium underaccumulation 1
Chromosome Location
Chr.4: 15132011 - 15136696 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119262.6 NP_194841.2 18 - 19 1589 114
NM_202918.2 NP_974647.1 18 - 19 1530 114

View Full Product Details