Assay Design



Gene Transcripts

Gene Symbol
MRE11
Entrez Gene ID
835514
Gene Name
DNA repair and meiosis protein (Mre11)
Gene Aliases
ARABIDOPSIS MEIOTIC RECOMBINATION 11, AT5G54260, ATMRE11, MDK4.8, MDK4_8, MEIOTIC RECOMBINATION 11
Chromosome Location
Chr.5: 22032038 - 22038052 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345081.1 NP_001330550.1 17 - 18 1579 74
NM_124806.2 NP_200237.1 19 - 20 1765 74
GenBank mRNA AJ243822.1 - 19 - 20 1734 74

View Full Product Details