Assay Design



Gene Transcripts

Gene Symbol
G18F
Entrez Gene ID
835562
Gene Name
autophagy 18 F-like protein
Gene Aliases
ARABIDOPSIS THALIANA HOMOLOG OF YEAST AUTOPHAGY 18 (ATG18) F, AT5G54730, ATATG18F, ATG18F, K5F14.9, K5F14_9, homolog of yeast autophagy 18 (ATG18) F
Chromosome Location
Chr.5: 22233789 - 22237370 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001345098.1 NP_001332742.1 2 - 3 837 113
NM_124854.4 NP_200284.1 3 - 4 1540 113

View Full Product Details