Assay Design
Gene Transcripts
- Gene Symbol
- PPX2
- Entrez Gene ID
-
835619
- Gene Name
- protein phosphatase X 2
- Gene Aliases
-
AT5G55260, MCO15.21, MCO15_21, PPX-2, PROTEIN PHOSPHATASE X -2, protein phosphatase X 2
- Chromosome Location
-
Chr.5: 22416578 - 22418990 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_124908.4 |
NP_200337.1 |
2 - 3 |
522 |
111 |
GenBank mRNA |
BT024716.1 |
- |
2 - 3 |
294 |
111 |
View Full Product Details