Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001203614.1 | NP_001190543.1 | 11 - 12 | 2389 | 95 |
NM_001203615.1 | NP_001190544.1 | 5 - 6 | 1326 | 95 | |
NM_124912.5 | NP_200341.1 | 11 - 12 | 2401 | 95 | |
GenBank mRNA | X57544.1 | - | - | 2326 | 95 |