Assay Design



Gene Transcripts

Gene Symbol
CRU3
Entrez Gene ID
828970
Gene Name
cruciferin 3
Gene Aliases
AT4G28520, CRC, CRUCIFERIN C, F20O9.210, F20O9_210, cruciferin 3
Chromosome Location
Chr.4: 14087556 - 14089796 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001084990.1 NP_001078459.1 1 - 2 336 78
NM_001341920.1 NP_001328588.1 1 - 2 336 78
NM_118994.4 NP_194581.1 1 - 2 347 78
NM_179133.2 NP_849464.1 1 - 2 343 78
NM_179134.1 NP_849465.1 1 - 2 336 78

View Full Product Details