Assay Design



Gene Transcripts

Gene Symbol
IMPA-4
Entrez Gene ID
837448
Gene Name
importin alpha
Gene Aliases
AT1G09270, T12M4.2, T12M4_2, importin alpha isoform 4
Chromosome Location
Chr.1: 2994331 - 2998222 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_100797.6 NP_172398.1 1 - 2 400 110
NM_179292.3 NP_849623.1 1 - 2 400 110
GenBank mRNA AK316689.1 - 1 - 2 297 110
AY090944.1 - 1 - 2 323 110
AY117313.1 - 1 - 2 225 110

View Full Product Details