Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035934.2 | NP_001031011.1 | 6 - 7 | 988 | 62 |
NM_100797.6 | NP_172398.1 | 6 - 7 | 1075 | 62 | |
NM_179292.3 | NP_849623.1 | 6 - 7 | 1075 | 62 | |
GenBank mRNA | AK316689.1 | - | 6 - 7 | 972 | 62 |
AY090944.1 | - | 6 - 7 | 998 | 62 | |
AY117313.1 | - | 6 - 7 | 900 | 62 |