Assay Design



Gene Transcripts

Gene Symbol
ACA1
Entrez Gene ID
839670
Gene Name
autoinhibited Ca2+-ATPase 1
Gene Aliases
AT1G27770, CA2+-ATPASE, PEA1, PLASTID ENVELOPE ATPASE 1, T22C5.23, T22C5_23, autoinhibited Ca2+-ATPase 1
Chromosome Location
Chr.1: 9671632 - 9676307 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332787.1 NP_001321522.1 3 - 4 402 94
NM_102542.1 NP_564295.1 2 - 3 384 94
NM_179385.2 NP_849716.1 2 - 3 433 94

View Full Product Details