Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001332786.1 | NP_001321521.1 | 2 - 3 | 2408 | 73 |
NM_001332787.1 | NP_001321522.1 | 4 - 5 | 2347 | 73 | |
NM_102542.1 | NP_564295.1 | 4 - 5 | 2107 | 73 | |
NM_179385.2 | NP_849716.1 | 3 - 4 | 2378 | 73 |