Assay Design



Gene Transcripts

Gene Symbol
ML5
Entrez Gene ID
839816
Gene Name
MEI2-like protein 5
Gene Aliases
AML5, AT1G29400, F15D2.30, F15D2_30, MEI2-like protein 5
Chromosome Location
Chr.1: 10289998 - 10294630 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001332851.1 NP_001320501.1 6 - 7 1510 113
NM_102680.2 NP_174233.2 6 - 7 1118 113
NM_179396.3 NP_849727.1 6 - 7 1267 113
GenBank mRNA AY062613.1 - 6 - 7 1154 113
AY070368.1 - 6 - 7 1157 113
AY091327.1 - 5 - 6 994 113

View Full Product Details