Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001332851.1 | NP_001320501.1 | 7 - 8 | 1605 | 65 |
NM_102680.2 | NP_174233.2 | 7 - 8 | 1213 | 65 | |
NM_179396.3 | NP_849727.1 | 7 - 8 | 1362 | 65 | |
GenBank mRNA | AY062613.1 | - | 7 - 8 | 1249 | 65 |
AY070368.1 | - | 7 - 8 | 1252 | 65 | |
AY091327.1 | - | 6 - 7 | 1089 | 65 |