Assay Design



Gene Transcripts

Gene Symbol
CAT2
Entrez Gene ID
842170
Gene Name
cationic amino acid transporter 2
Gene Aliases
AT1G58030, cationic amino acid transporter 2
Chromosome Location
Chr.1: 21464001 - 21468505 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_179491.3 NP_849822.1 11 - 12 1924 73
GenBank mRNA AF367304.1 - 11 - 12 1829 73
BT046174.1 - 11 - 12 1560 73

View Full Product Details