Assay Design



Gene Transcripts

Gene Symbol
ALIS1
Entrez Gene ID
820456
Gene Name
ALA-interacting subunit 1
Gene Aliases
ALA-interacting subunit 1, AT3G12740
Chromosome Location
Chr.3: 4049309 - 4051775 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_112110.4 NP_566435.1 2 - 3 578 112
GenBank mRNA AF410309.1 - 2 - 3 207 112
AY045904.1 - 2 - 3 283 112
AY087607.1 - 2 - 3 372 112
AY091210.1 - 2 - 3 148 112

View Full Product Details