Assay Design
Gene Transcripts
- Gene Symbol
- MAPKKK7
- Entrez Gene ID
-
820555
- Gene Name
- mitogen-activated protein kinase kinase kinase 7
- Gene Aliases
-
AT3G13530, MAP3K EPSILON PROTEIN KINASE, MAP3KE1, mitogen-activated protein kinase kinase kinase 7
- Chromosome Location
-
Chr.3: 4411688 - 4423000 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_112199.3 |
NP_187962.1 |
10 - 11 |
4551 |
66 |
View Full Product Details