Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001338114.1 | NP_001326872.1 | 5 - 6 | 991 | 70 |
NM_112278.3 | NP_188037.1 | 4 - 5 | 1333 | 70 | |
GenBank mRNA | AK221834.1 | - | 4 - 5 | 909 | 70 |
AY054230.1 | - | 4 - 5 | 887 | 70 | |
AY062690.1 | - | 4 - 5 | 889 | 70 | |
BT002522.1 | - | 4 - 5 | 889 | 70 | |
BT008484.1 | - | 4 - 5 | 846 | 70 |