Assay Design



Gene Transcripts

Gene Symbol
RSZ22
Entrez Gene ID
829285
Gene Name
serine/arginine-rich 22
Gene Aliases
AT4G31580, At-RSZ22, AtRSZ22, F28M20.230, F28M20_230, RS-CONTAINING ZINC FINGER PROTEIN 22, RS-containing zinc finger protein 22, RSZP22, SRZ-22, SRZ22, serine/arginine-rich 22
Chromosome Location
Chr.4: 15306544 - 15309035 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085005.1 NP_001078474.1 3 - 4 255 59
NM_119307.4 NP_194886.1 3 - 4 409 59
GenBank mRNA AF033586.1 - 3 - 4 222 59
AY065387.1 - 3 - 4 249 59
AY086525.1 - 3 - 4 274 59
AY117206.1 - 2 - 3 173 59

View Full Product Details