Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001085005.1 | NP_001078474.1 | 4 - 5 | 513 | 79 |
NM_119307.4 | NP_194886.1 | 4 - 5 | 667 | 79 | |
GenBank mRNA | AF033586.1 | - | 4 - 5 | 480 | 79 |
AY065387.1 | - | 4 - 5 | 507 | 79 | |
AY086525.1 | - | 4 - 5 | 532 | 79 | |
AY117206.1 | - | 3 - 4 | 431 | 79 |