Assay Design



Gene Transcripts

Gene Symbol
CESA1
Entrez Gene ID
829376
Gene Name
cellulose synthase 1
Gene Aliases
ANY1, AT4G32410, AtCESA1, CELLULOSE SYNTHASE 1, F8B4.110, F8B4_110, RADIALLY SWOLLEN 1, RSW1, anisotropy1, cellulose synthase 1
Chromosome Location
Chr.4: 15640490 - 15646844 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119393.3 NP_194967.1 13 - 14 3118 89
GenBank mRNA AK226243.1 - 13 - 14 2797 89
BT008654.1 - 13 - 14 2806 89
KJ138727.1 - 13 - 14 2662 89

View Full Product Details