Assay Design



Gene Transcripts

Gene Symbol
YUC1
Entrez Gene ID
829389
Gene Name
Flavin-binding monooxygenase family protein
Gene Aliases
AT4G32540, L23H3.20, L23H3_20, YUC, YUCCA, YUCCA 1
Chromosome Location
Chr.4: 15700829 - 15703000 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001342156.1 NP_001329230.1 3 - 4 1020 69
NM_119406.3 NP_194980.1 3 - 4 1089 69

View Full Product Details