Assay Design



Gene Transcripts

Gene Symbol
KAT3
Entrez Gene ID
829400
Gene Name
potassium channel protein
Gene Aliases
A. thaliana low-K+-tolerant 1, ARABIDOPSIS THALIANA K+ RECTIFYING CHANNEL 1, AT4G32650, ATKC1, AtLKT1, F4D11.150, F4D11_150, K+ INWARD RECTIFYING CHANNEL PROTEIN, KC1, potassium channel in Arabidopsis thaliana 3
Chromosome Location
Chr.4: 15751271 - 15755047 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036696.1 NP_001031773.1 12 - 13 1934 101
NM_119417.4 NP_194991.1 12 - 13 2139 101
GenBank mRNA AY052233.1 - 12 - 13 1937 101
BT003093.1 - 12 - 13 1889 101
Z83202.1 - - 1935 101

View Full Product Details