Assay Design



Gene Transcripts

Gene Symbol
KAT3
Entrez Gene ID
829400
Gene Name
potassium channel protein
Gene Aliases
A. thaliana low-K+-tolerant 1, ARABIDOPSIS THALIANA K+ RECTIFYING CHANNEL 1, AT4G32650, ATKC1, AtLKT1, F4D11.150, F4D11_150, K+ INWARD RECTIFYING CHANNEL PROTEIN, KC1, potassium channel in Arabidopsis thaliana 3
Chromosome Location
Chr.4: 15751271 - 15755047 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001036696.1 NP_001031773.1 6 - 7 1300 100
NM_119417.4 NP_194991.1 6 - 7 1502 100
NM_202936.1 NP_974665.1 6 - 7 1309 100
GenBank mRNA AY052233.1 - 6 - 7 1300 100
BT003093.1 - 6 - 7 1252 100
U81239.1 - 6 - 7 1257 100
Z83202.1 - - 1298 100

View Full Product Details