Assay Design



Gene Transcripts

Gene Symbol
PFK6
Entrez Gene ID
829420
Gene Name
phosphofructokinase 6
Gene Aliases
AT4G32840, T16I18.50, T16I18_50, phosphofructokinase 6
Chromosome Location
Chr.4: 15844576 - 15848552 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119437.3 NP_195010.1 8 - 9 919 79
GenBank mRNA AK227471.1 - 8 - 9 808 79
BT006001.1 - 8 - 9 672 79

View Full Product Details