Assay Design



Gene Transcripts

Gene Symbol
APM1
Entrez Gene ID
829446
Gene Name
aminopeptidase M1
Gene Aliases
AMINOPEPTIDASE M1, AT4G33090, ATAPM1, F4I10.20, F4I10_20, aminopeptidase M1
Chromosome Location
Chr.4: 15965593 - 15970557 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119463.5 NP_195035.2 3 - 4 604 87
GenBank mRNA AK227105.1 - 3 - 4 501 87
AY064961.1 - 3 - 4 502 87
AY143574.1 - 3 - 4 477 87
BT005811.1 - 3 - 4 465 87

View Full Product Details