Assay Design



Gene Transcripts

Gene Symbol
APM1
Entrez Gene ID
829446
Gene Name
aminopeptidase M1
Gene Aliases
AMINOPEPTIDASE M1, AT4G33090, ATAPM1, F4I10.20, F4I10_20, aminopeptidase M1
Chromosome Location
Chr.4: 15965593 - 15970557 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119463.5 NP_195035.2 7 - 8 1150 84
GenBank mRNA AK227105.1 - 7 - 8 1047 84
AY064961.1 - 7 - 8 1048 84
AY143574.1 - 7 - 8 1023 84
BT005811.1 - 7 - 8 1011 84

View Full Product Details