Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001342233.1 | NP_001329220.1 | 9 - 10 | 1571 | 98 |
NM_119537.7 | NP_567936.5 | 9 - 10 | 1391 | 98 | |
GenBank mRNA | AY057657.1 | - | 9 - 10 | 1377 | 98 |
AY070065.1 | - | 9 - 10 | 1379 | 98 |