Assay Design



Gene Transcripts

Gene Symbol
DML3
Entrez Gene ID
829552
Gene Name
demeter-like protein 3
Gene Aliases
AT4G34060, F28A23.180, F28A23_180, demeter-like protein 3
Chromosome Location
Chr.4: 16313366 - 16319426 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001125640.1 NP_001119112.1 8 - 9 1740 70
NM_001342250.1 NP_001329149.1 9 - 10 1967 70
NM_001342251.1 NP_001329150.1 6 - 7 1992 70
NM_001342252.1 NP_001329148.1 7 - 8 1840 70
NM_119567.4 NP_195132.3 8 - 9 1894 70

View Full Product Details