Assay Design



Gene Transcripts

Gene Symbol
LEJ1
Entrez Gene ID
829558
Gene Name
Cystathionine beta-synthase (CBS) family protein
Gene Aliases
AT4G34120, CBS domain containing protein 2, CBSX2, CDCP1, CYSTATHIONE [BETA]-SYNTHASE DOMAIN-CONTAINING PROTEIN 1, F28A23.120, F28A23_120, LOSS OF THE TIMING OF ET AND JA BIOSYNTHESIS 1
Chromosome Location
Chr.4: 16340907 - 16343118 on Build TAIR10.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_119573.5 NP_567952.1 3 - 4 660 74
GenBank mRNA AF360331.1 - 3 - 4 428 74
AY056339.1 - 3 - 4 373 74

View Full Product Details