Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001342297.1 | NP_001329728.1 | 12 - 13 | 1223 | 96 |
NM_119630.4 | NP_195190.1 | 12 - 13 | 1469 | 96 | |
GenBank mRNA | AY099868.1 | - | 12 - 13 | 1272 | 96 |
BT003419.1 | - | 12 - 13 | 1169 | 96 | |
D29017.1 | - | 12 - 13 | 1383 | 96 | |
U79159.1 | - | 12 - 13 | 1296 | 96 | |
X86692.1 | - | - | 1282 | 96 |