Assay Design
Gene Transcripts
- Gene Symbol
- MRL1
- Entrez Gene ID
-
829635
- Gene Name
- Pentatricopeptide repeat (PPR) superfamily protein
- Gene Aliases
-
AT4G34830, F11I11.70, F11I11_70, MATURATION OF RBCL 1, PDE346, PIGMENT DEFECTIVE 346
- Chromosome Location
-
Chr.4: 16599597 - 16606135 on Build TAIR10.1
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_119649.5 |
NP_195209.2 |
1 - 2 |
467 |
101 |
View Full Product Details